Would you like to log in again?
MAT BLUE TABLE PREMIUM 2X4' EA
Supplier: DESCO
Mat^ SCC* Premium three layer vinyl mat offers outstanding ESD protection and value. 0.125 inch thick three layer construction provides excellent electrical properties and is compatible with constant monitors. Rtg: 10-7 ohms Rtt: 10-7 ohms. 2' x 4' blue mat wi
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
MAT BLUE TABLE PREMIUM 2X4' EA 89024-596 TM2448L3BL-L Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty