Would you like to log in again?
SHOE GROUNDING HEEL STRAP DISP BLACK/ YL
Supplier: DESCO
Foot Grounder Disposable Yellow/Black 12in^ Foot Grounder Disposable, Highly Visible Yellow Polyester Fabric, Non-Marring Material, Adhesive Holds Grounder to Sole of Shoe, Meets ANSI/ESD S20.20 Foot Grounder required limits tested per ESD SP9.2 and ESD TR53, Colour: Yellow/Black, Length: 12in
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
SHOE GROUNDING HEEL STRAP DISP BLACK/ YL 89024-651 5402 Each Retrieving
Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty