Would you like to log in again?
LABEL LEADFREE 175X175IN 500RL
Supplier: DESCO
Labels^ (ROHSLABEL) SCC* ROHS labels mark your lead free products, or to designate equipment and materials that are used for lead free production. This green 1.75 x 1.75 inch adhesive label shows the inch PB free inch symbol. 500 labels per 3 inch paper core.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
LABEL LEADFREE 175X175IN 500RL 89024-660 ROHSLABEL Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty