Would you like to log in again?
Ribbon Utility Scissors, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
47-1146 47-1247
89024-950EA 99.3 USD InStock
89024-950 89024-940
Ribbon Utility Scissors, OR Grade, Sklar
Scissors
Ribbon Utility Scissors are a multipurpose instrument used for cutting lightweight, non-sterile materials such as gauze, bandages and dressing material.
This product is straight with smooth edges, and ribbon style handles for additional comfort.
Angular Scissors are finger ring scissors comparable to iris scissors.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty