Would you like to log in again?
Adson Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
47-1450
89024-968EA 118.73 USD InStock
89024-968
Adson Tissue Forceps, OR Grade, Sklar
Forceps
Adson Tissue Forceps are for holding and manipulating delicate tissues.
These forceps have a wide, flat thumb grasp area that is commonly serrated. The jaws are short and the tips are narrow.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty