Would you like to log in again?
Ferris Smith Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
47-1470
89024-970EA 242.62 USD InStock
89024-970
Ferris Smith Tissue Forceps, OR Grade, Sklar
Forceps
Ferris Smith Tissue Forceps are heavyweight forceps used with fibrous tissue, commonly in colorectal procedures
Censitrac Ready products are industry standard and constructed of stainless steel.
These thumb forceps feature a wide section of the handle for greater control handling firm tissue. They also have slightly wider tips that are available with 1 X 2 or 2 X 3 teeth for a more secure grasp of the tissue. This product is straight with cross-serrated tips, 1x2 teeth.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty