Would you like to log in again?
Guthrie Fixation Hook, OR Grade, Sklar®
Supplier: Sklar
47-2651 47-2650
89025-020EA 135.38 USD InStock
89025-020 89025-022
Guthrie Fixation Hook, OR Grade, Sklar®
Retractors
Sklar's® Guthrie Hook is a double-prong tissue hook used for handling and retracting tissue
They are often used in dermatologic and plastic surgery procedures but the tiny hooks also make this retractor useful in ophthalmologic and intranasal procedures. It is straight with two sharp, hooked, prong tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty