Would you like to log in again?
Hoff Towel Clamp, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
47-3057 47-3040
89025-042EA 195.51 USD InStock
89025-042 89025-044
Hoff Towel Clamp, OR Grade, Sklar®
Forceps
Sklar® Hoff Towel Clamps are a non-perforating clamp used to secure instruments and towels to the drape surrounding a surgical site
The jaws and tips are curved while the shanks and handle are flat so they can rest on the patient. The tip of this towel clamp are round with a cross-serrated surface so they grip tightly but do not penetrate the surgical drape. They also have ratcheted finger ring handles so a firm hold can be secured. This clamp should never be used on skin.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty