Would you like to log in again?
Cushing Brain Forceps, OR Grade, Sklar
Supplier: Sklar
22-6870 50-2670 50-2970 50-2870 22-6770
89025-070EA 91.23 USD InStock
89025-070 89025-390 89025-392 89025-068
Cushing Brain Forceps, OR Grade, Sklar
Forceps
Sklar® Cushing forceps are light to medium weight thumb forceps used on delicate tissue. They are commonly used in neurosurgery. Cushing forceps have very narrow tips and are useful in small surgical areas.
They securely grasp the tissue, but also have a stop peg to prevent an overly harsh grasp that may crush the tissue
Censitrac Ready products are industry standard and constructed of stainless steel.
They are available with a standard, Gutch, or bayonet handle depending on the surgical site and user preference.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty