Would you like to log in again?
Adson Bayonet Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
50-3371
89025-080EA 141.86 USD InStock
89025-080
Adson Bayonet Dressing Forceps, OR Grade, Sklar
Forceps
Censitrac Ready products are industry standard and constructed of stainless steel.
  • Sklar® Adson Bayonet Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds. They have a wide thumb grasp for increased precision and control. The bayonet style handle allows these forceps to be used in areas with a compromised view.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty