Would you like to log in again?
TC Adson Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
51-3049
89025-096EA 318.74 USD InStock
89025-096
TC Adson Tissue Forceps, OR Grade, Sklar
Forceps
TC Adson Tissue Forceps are for holding and manipulating delicate tissues.
Sklar permanently bonds tungsten carbide inserts to the instrument, which provides maximum control and precision for the user, as well as increasing the length of time the instrument will remain sharp. These forceps have a wide, flat thumb grasp area that is commonly slightly grooved for an improved grip. The jaws are short and the tips are narrow. These TC Adson tissue forceps are identified by gold handles.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty