Would you like to log in again?
Potts-Smith TC Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
51-3460 51-3480
89025-102EA 358.65 USD InStock
89025-102 89025-100
Potts-Smith TC Dressing Forceps, OR Grade, Sklar
Forceps
Sklar® Potts-Smith TC Dressing Forceps are most commonly used for application and changing of dressings. They may also be used for dissection of fine or single-layer tissue.
Cross-serrated tips provide a solid grip on both materials and tissue
Censitrac Ready products are industry standard and constructed of stainless steel.
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty