Would you like to log in again?
Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 
DeBakey Bulldog Clamp, OR Grade, Sklar®
Supplier: Sklar
52-1530 52-1542 52-1541 52-1540 52-1531 52-1543 52-1533
89025-136EA 360.07 USD InStock
89025-136 89025-138 89025-140 89025-130 89025-142 89025-132 89025-144
DeBakey Bulldog Clamp, OR Grade, Sklar®
Forceps
Sklar's® DeBakey Atraumatic Bulldog Clamp is a small cross-action thumb clamp used for halting blood flow in a vessel
They are most commonly used in cardiovascular or cardiothoracic procedures. They are available curved or straight, in multiple lengths
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty