Would you like to log in again?
DeBakey Atraumatic Forceps, OR Grade, Sklar
Supplier: Sklar
52-5262 52-5198 52-5202 52-5062 52-5162 52-5095 52-5277 52-5177 52-5098 52-5077 52-5195 52-5295 52-5312 52-5309 52-5297
89025-158EA 505.86 USD InStock
89025-158 10196-450 89025-184 89025-162 89025-174 89025-196 89025-186 89025-176 89025-154 89025-198 89025-166 89025-188 89025-156 89025-190 89025-170
DeBakey Atraumatic Forceps, OR Grade, Sklar
Forceps
DeBakey Atraumatic Tissue Forceps are a versatile thumb forceps used for grasping fine tissue. They are commonly used in vascular and general surgery.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty