Would you like to log in again?
Operating Scissors, Sklar
Supplier: Sklar
15-2055 22-1545 14-1065 13-1060 14-2060 15-1055 22-1550 22-1445 14-2065 14-1055 22-1255 13-2050 22-1655 13-1055 22-1355 15-1060 15-1050 13-2045 14-1072 13-2060 14-2055 22-1350 22-1145 22-1645 13-1045 14-1045 15-2050 13-1065 22-1250 15-2045 14-1060 22-1245 13-1072 22-1155 14-1050 13-2055 13-2065 15-2065 22-1455 14-2045 22-1555 22-1450 15-2060 15-1045 14-2050 22-1150 22-1650 13-2072 13-1050 22-1345 15-1065
82029-922EA 70.8 USD InStock
82029-922 82029-888 82029-924 89025-236 82029-926 89025-238 82029-928 89025-274 82029-884 89025-276 82029-920 82029-886 82029-890 89025-246 82029-910 82029-954 82029-912 82029-956 82029-914 82029-916 89025-240 82029-950 82029-952 89025-244 82029-918 82029-900 89025-256 82029-902 89025-214 89025-258 82029-948 82029-904 89025-216 89025-218 82029-906 89025-250 82029-940 89025-254 10195-204 82029-908 10195-208 10195-206 82029-932 82029-934 82029-936 82029-938 82029-892 82029-894 82029-896 89025-264 82029-898
Operating Scissors, Sklar
Scissors
Operating Scissors, also known as Surgical Scissors, are a versatile surgical instrument.
With three available tip configurations (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures, and multiple lengths, these scissors are used for general, multi-purpose cutting, and dissecting
Censitrac Ready OR Grade Operating Scissors are industry standard and constructed of stainless steel.
Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty