Would you like to log in again?
Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
22-2460 22-2450
89025-282EA 168.65 USD InStock
89025-282 89025-284
Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Scissors
Metzenbaum Dissecting Scissors are most commonly used for dissecting and cutting tissue.
They are not recommended for cutting sutures, drains, or heavy tissue.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty