Would you like to log in again?
Ragnell Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
22-2657 22-2653 22-2652
89025-300EA 164.22 USD InStock
89025-300 76192-744 89025-298
Ragnell Dissecting Scissors, OR Grade, Sklar®
Scissors
Sklar's® Ragnell Scissors, also known as Kilner/Ragnell scissors, are finger ring scissors
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Long shanks and short jaws allow the user greater control with less effort during dissection. This combined with flat blades and blunt tips often make Ragnell scissors the dissection instrument of choice during longer procedures.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty