Would you like to log in again?
Sarot Needle Holder, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
22-5050 20-4170 20-3910
10195-174EA 328.66 USD InStock
10195-174 10195-172 89025-376
Sarot Needle Holder, OR Grade, Sklar
Forceps
Sklar's® Sarot Needle Holder is used for approximation suturing, often in cardiovascular procedures.
It is a ratcheted, finger ring instrument that is available in long, short, and pediatric lengths
Censitrac Ready OR Grade Sarot Needle Holders are industry standard and constructed of stainless steel.
The shanks curve inward slightly near the finger rings, then extend straight out to short jaws with tips that feature cross-serrations and one central groove. The cross-serrated surface and groove ensure a non-slip hold on the needle, while the ratcheted locking mechanism provides additional security of the hold.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty