Would you like to log in again?
Allen Intestinal Clamp, OR Grade, Sklar
Supplier: Sklar
22-6160
89025-382EA 336.64 USD InStock
89025-382
Allen Intestinal Clamp, OR Grade, Sklar
Forceps
Sklar's® Allen Intestinal Clamp is a ratcheted, finger ring clamp primarily used for grasping tissue or hemostasis
It is very similar to a toothed Kocher clamp, but is more lightweight. The jaws of the Allen clamp are longitudinally serrated with 1x2 teeth. This product is straight with an overall length of 6 inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty