Would you like to log in again?
Bandage Scissors, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
11-1252
89025-446EA 164.86 USD InStock
89025-446
Bandage Scissors, OR Grade, Sklar
Scissors
Sklar's® Hi-Level Bandage Scissors are primarily used for removing tight bandages.
The tip of the lower blade features a flattened blunt nodule that is intended to slide between bandages and skin without harming the skin
Censitrac Ready products are industry standard and constructed of stainless steel.
This product has one sharp tip along with the nodule tip. The shanks are strongly curved before the screw lock and the blades are straight.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty