Would you like to log in again?
Hercules Utility Scissors, OR Grade, Sklar
Supplier: Sklar
11-1297
89025-466EA 307.31 USD InStock
89025-466
Hercules Utility Scissors, OR Grade, Sklar
Scissors
Hercules Utility Scissors are heavy scissors used for cutting heavy bandage material, aluminum, and plaster casts.
This product is angled with one serrated and one smooth edge, blunt/blunt tips, and a length of 7-1/2 inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty