Would you like to log in again?
Deaver Scissors, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
15-1155 14-1155 13-1155 13-2155 14-2155
89025-488EA 92 USD InStock
89025-488 89025-490 89025-480 89025-482 89025-486
Deaver Scissors, OR Grade, Sklar
Scissors
Deaver Operating Scissors have all the functionality of standard operating scissors, but also feature a delicate pattern. Three tip configurations are available (sharp/sharp, blunt/blunt, or sharp/blunt), varying curvatures and multiple lengths, allowing these scissors to be used for multipurpose cutting and dissecting.
Sharp Operating Scissors cut tougher tissue and are frequently used for dissecting, whereas blunt Operating Scissors are more often used for cutting materials such as sutures, tubing, or gauze.
Operating room grade instruments are made from the highest grade surgical steel and incorporate the finest level of craftsmanship. Superior manufacturing and quality inspection guarantee that Sklar® instruments meet the demands of customers. These instruments have been designed to provide exceptional comfort and precision all while ensuring the durability and strength that has been provided.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty