Would you like to log in again?
Stitch Scissors, OR Grade, Sklar
Supplier: Sklar
15-3150
89025-536EA 153.62 USD InStock
89025-536
Stitch Scissors, OR Grade, Sklar
Scissors
Stitch Scissors are used for suture removal.
These scissors have a small hook shaped tip on one blade that slides under sutures to slightly lift them before cutting for removal. This hook also holds the suture so it does not slip off the blade of the scissors prior to cutting. Stitch scissors should not be used for cutting tissue.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty