Would you like to log in again?
Sklarhone™ Mayo Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
15-3331 15-3426 15-3330 15-3326 15-3325
89025-554EA 262.32 USD InStock
89025-554 89025-556 89025-558 89025-582 89025-552
Sklarhone™ Mayo Dissecting Scissors, OR Grade, Sklar®
Scissors
Sklarhone™ Mayo Dissecting Scissors are a variation of standard Mayo Scissors that feature one knife-like razor sharp edge and one standard scissors edge
This allows the user to make the sharp incisions of a knife with the cutting ability of scissors, while catching skin and pulls are minimized. These scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty