Would you like to log in again?
Supercut Sklarhone™ Iris scissors, OR Grade, Sklar
Supplier: Sklar
15-3457 15-3452
89025-594EA 258.98 USD InStock
89025-594 89025-596
Supercut Sklarhone™ Iris scissors, OR Grade, Sklar
Scissors
Supercut Sklarhone™ Iris scissors combine one knife-like razor edge and one micro-serrated scissor edge.
These scissors can be used to cut both delicate and heavy tissue because of their unique design. The micro-serrations of the Supercut Sklarhone™ scissors minimize tearing of tissue so cuts and edges are smooth and clean. Originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal. In addition to ophthalmic procedures, Iris Scissors are now often used in emergency rooms and for OB/GYN and dermatologic procedures
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty