Would you like to log in again?
Sklarcut™ Mayo Dissecting Scissors, OR Grade, Sklar®
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sklarcut™
15-3560 15-3568 15-3555 15-3566 15-3565 15-3550
76192-020EA 265.87 USD InStock
76192-020 89025-634 89025-636 89025-638 10193-388 89025-640
Sklarcut™ Mayo Dissecting Scissors, OR Grade, Sklar®
Scissors
Sklarcut™ Mayo Dissecting Scissors are a variation of standard Mayo Scissors that feature ultra-fine serrations on one blade to reduce trauma when cutting both delicate and course tissue
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
These serrations assist in securely holding the tissue during cutting and minimizes tissue from slipping so the most accurate and clean cuts can be made. These scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference. Sklarcut™ scissors are identified by a single gold coated finger ring for the serrated blade.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty