Would you like to log in again?
Sklar Edge™ Tungsten Carbide Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Sklar Edge™
16-3513 16-3526 16-1815 16-1805 16-3527 16-2005 16-1915 16-2105 16-1905 16-3506 16-2115 16-3503 16-3514
89025-698EA 498.53 USD InStock
89025-698 89025-688 76191-988 89025-690 89025-680 76192-018 76192-048 76192-016 89025-694 76192-014 89025-684 76192-062 76192-060
Sklar Edge™ Tungsten Carbide Metzenbaum-Lahey Dissecting Scissors, OR Grade, Sklar
Scissors
Sklar Edge™ Tungsten Carbide Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue.
  • For more than 120 years, Sklar has set the standard for surgical instrumentation
  • With five major surgical instruments lines, Sklar is able to deliver superior precision for any surgical procedure
  • All Sklar products are industry standard
Metzenbaum-Lahey scissors tend to be shorter than traditional Metzenbaum scissors.
Sklar Edge™ Tungsten Carbide Scissors provide an exceptionally durable and hard cutting edge. These scissors are designed to stay sharp for a significantly longer period of time than standard scissors. Sklar permanently bonds tungsten carbide inserts to the blades of the scissors which maximizes control and precision. Sklar Edge™ scissors can be identified by two gold coated finger rings.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty