Would you like to log in again?
Sklar Edge™ Tungsten Carbide Strabismus Scissors, OR Grade, Sklar
Supplier: Sklar
16-2605 16-2705
89025-722EA 367.72 USD InStock
89025-722 89025-726
Sklar Edge™ Tungsten Carbide Strabismus Scissors, OR Grade, Sklar
Scissors
Sklar Edge™ Tungsten Carbide Strabismus Scissors provide an exceptionally durable and hard cutting edge
TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.
They are an ophthalmic dissecting instrument and are used during procedures for the treatment of Strabismus, otherwise known as "cross-eyes."
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty