Would you like to log in again?
Econo™ Sterile Adson Dressing Forceps, Floor Grade, Sklar
Supplier: Sklar
Econo™
96-2577 96-2571 96-2655 96-2570
89025-740CS 433.34 USD InStock
89025-740 89023-968 89023-886 89023-888
Econo™ Sterile Adson Dressing Forceps, Floor Grade, Sklar
Forceps
Econo™ Sterile Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
  • Wide thumb grasp for increased precision and control
  • Serrated tips
  • Ready-to-use
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments sold in bulk quantities. This product is straight with serrated tips.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty