Would you like to log in again?
Pipette Tips for AP96, FX/NX, 1000, 2000, and 3000 Series Workstations, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
BioRobotix®
918-011 915-262 919-021G 141-96RS 917-262 142-96RS 912-021 919-011 142-96R 915-261 918-261-05 935-021 918-252 141-96R 916-261 916-262 919-021-05 917-261 912-262 919-252 916-021
72830-528CS 1851.39 USD InStock
72830-528 89025-876 72830-408 72830-518 89025-878 72830-216 72830-218 72830-212 89025-880 72830-412 72830-210 72830-353 46300-372 46300-362 46300-370 490014-540 46300-360 76457-542 72830-290 53226-018 53226-016
Pipette Tips for AP96, FX/NX, 1000, 2000, and 3000 Series Workstations, Thermo Scientific
Pipette Tips
Molecular BioProducts pipette tips are designed to fit AP96, FX/NX, 1000, 2000, and 3000 liquid handling workstations.
  • Designed for compatibility and performance
  • Variety of sizes and styles available
  • Lot-tested using 15-point program
BioRobotix® robotic pipette tips are available in low retention for critical samples and in extended length for increased volume and the ability to reach into deep well plates.
Genomic tips are excellent for pipetting of more viscous samples. The tips have an orifice that is up to 260% larger than the standard workstation tip orifice. This provides more precision when pipetting viscous tissue samples, such as blood, and prevents shearing of genomic DNA during sample preparation and processing.
Automation pipette tips for the 1000 and 2000 automated liquid handling systems feature precise design and optimal fit for accuracy. Wide selection of tips in both standard and liquid-level sensing carbon black-conductive polypropylene are available. Automation pipette tips for the 1000 and 2000 are certified RNase-, DNase-, and pyrogen-free.
Automation pipette tips for the FX and NX automated liquid handling systems have been designed, validated, and tested on the FX workstation to ensure equal performance without the need to modify your existing protocols and programs. The low percentage Coefficient of Variation (%CV) ensures product consistency and performance from every lot. Automation pipette tips for the FX and NX are certified RNase, DNase, DNA, endotoxin, bioburden, and pyrogen free.
The pipette tips, racks and lids reduce static charges so tips pick up straight and eject perfectly each time. These tips are designed with SoftFit™ feature that enhances workstation performance and longevity by giving a more secure fit with reliable, consistent loading and ejecting. Filter tips utilize the ART barrier for guranteed protection against cross-contamination.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty