Would you like to log in again?
VWR® Funnels, PTFE
Supplier: VWR
VWR®
039.050 039.100 039.150 039.070 039.030
89026-144EA 76.89 USD InStock
89026-144 89026-142 89026-140 89026-148 89026-146
VWR® Funnels, PTFE
Funnels Filtration Funnels Filter Funnels
Isostatically molded, FDA approved pure PTFE molding provides a durable, heavy wall construction with a smooth, non-stick internal finish.
  • Chemically inert
  • May be used from 0 to 280 °C (32 to 536 °F)
Suitable for a variety of laboratory applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty