Would you like to log in again?
VWR® Magnetic Stir Bar Retrievers, PTFE
Supplier: VWR
VWR®
004.250 004.250.RE 004.350 004.450 004.150 004.450.RE 004.350.RE
89026-264EA 97.76 USD InStock
89026-264 89026-254 89026-262 89026-260 89026-258 89026-266 89026-256
VWR® Magnetic Stir Bar Retrievers, PTFE
Stirrers Magnetic Stirring Bars
Molded in pure, FDA-approved PTFE, these retrievers are inert and non-contaminating.
  • Inert
  • PTFE fabrication
  • Autoclavable
  • Magnetic cores available in different variants
Available with standard Alnico V or samarium cobalt rare earth magnetic core. Autoclavable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty