Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

VWR® Stirrer Blades with Plain Ends

Supplier: VWR
VWR®
002.1065.1 002.1125.1 002.1105.1 002.1075.1 002.1150.1
89026-276EA 20.98 USD InStock
89026-276 89026-274 89026-272 89026-270 89026-268
VWR® Stirrer Blades with Plain Ends
Stirrers Stirring Blades
Blades fit PTFE stirring shaft; hole diameter 6.5 mm, 3.2 mm thick.

  • For plain shafts
  • Very good chemical resistance
  • Inert
  • Temperature resistant up to +280°C

Ordering information: For use with 005.500 series plain stirrer shafts (sold separately). Contact your VWR representative for more information.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty