Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
VWR® Stirring Rods
Supplier: VWR
VWR®
003.250 003.100 003.400 003.200 003.300
89026-286PK 198.76 USD InStock
89026-286 89026-284 89026-282 89026-280 89026-290
VWR® Stirring Rods
Stirrers Stirring Rods
Available with a steel core or in solid PTFE, these rods are chemically resistant, inert, and non-abrasive.
  • Constructed from solid PTFE or with a steel core
  • Chemically resistant
  • Inert
  • Non-abrasive
Steel core rods have a rounded end, can be bent into a permanent shape, and are ideal when extra rigidity is required. Solid PTFE rods have a tapered end. Usable to 280°C (536°F).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty