Would you like to log in again?
VWR® Tweezers, PTFE
Supplier: VWR
VWR®
027.101 027.150 027.151 027.100
89026-418EA 100.93 USD InStock
89026-418 89026-426 89026-424 89026-420
VWR® Tweezers, PTFE
Forceps
Tweezers have a smooth PTFE non-stick surface.
  • PTFE construction
  • Non-contaminating
  • Inert
For use in temperatures of 0 to 280°C (32 to 540°F). Non-contaminating and inert to virtually all chemical reagents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty