Would you like to log in again?
EP Scientific Class 100 Low-Particle/Low Metal Bottles, HDPE, Thermo Scientific
Supplier: Thermo Scientific
156-125W/N/LP 150-01W/N/LP 157-250W/N/LP
89026-594CS 601.15 USD InStock
89026-594 89026-592 89026-596
EP Scientific Class 100 Low-Particle/Low Metal Bottles, HDPE, Thermo Scientific
Bottles Plastic Bottles
These bottles are ideal for break-resistant storage and packaging of ultrapure materials, off-line D.I.
  • Narrow-mouth, natural HDPE bottles
Bottles are certified to contain less than 20 particles per ml at >= 0.3 µ. The metals typically certified include Al, Ca, Cu, Fe, K, Mg, Mn, Na and Zn at <10 ppb.
Water and chemical sampling, testing for trace inorganic impurities, particle analysis, and for use in combination packaging.
Ordering information: Bottles are supplied with polypropylene caps and are double-bagged for extra cleanliness. Supplied with Certificate of Analysis.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty