Would you like to log in again?
EP Scientific Silanized Screw Thread Vials, Thermo Scientific
Supplier: Thermo Scientific
SAA-SV4-2 SAA-SV2-2 SCA-SV4-2
89026-618CS 346.77 USD InStock
89026-618 89026-616 89026-610
EP Scientific Silanized Screw Thread Vials, Thermo Scientific
Vials
These vials are mainly used to preserve the integrity of certain materials or extracts stored in glass containers.
  • Trace organic analysis
  • Storage of materials prone to adhering to glass
  • Extraction glassware
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty