Would you like to log in again?
O-RING KALREZ. 4079 SIZE 014
Supplier: Chemglass
Kalrez* 4079 Perfluroelastomer, Size:014^ Combines the elastomeric properties of Viton with the chemical resistance of PTFE, making it the best possible o-ring material available. Kalrez* 4079 should be considered for service in high temperature, corrosive environments. Size 014.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
O-RING KALREZ. 4079 SIZE 014 89027-186 CG-308K-014 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty