Would you like to log in again?
O-Rings, Perfluoro, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-309-137 CG-309-112 CG-309-212 CG-309-006 CG-309-111 CG-309-012 CG-309-008 CG-309-214 CG-309-019 CG-309-218 CG-309-115 CG-309-110 CG-309-018 CG-309-223 CG-309-226 CG-309-331 CG-309-217 CG-309-020 CG-309-011 CG-309-013 CG-309-206 CG-309-225 CG-309-109 CG-309-108 CG-309-121 CG-309-219 CG-309-220 CG-309-104 CG-309-201 CG-309-022 CG-309-027 CG-309-210 CG-309-107 CG-309-222 CG-309-024 CG-309-116 CG-309-243 CG-309-132 CG-309-009 CG-309-205 CG-309-007 CG-309-146 CG-309-229 CG-309-129 CG-309-122 CG-309-120 CG-309-114 CG-309-010 CG-309-117 CG-309-105 CG-309-136 CG-309-208 CG-309-204 CG-309-014
89027-284EA 53.1 USD InStock
89027-284 89027-286 89027-280 89027-282 89027-324 89027-248 89027-326 89029-332 89027-320 89027-288 89027-322 89027-318 89027-272 89027-274 89027-270 89027-314 89027-316 89027-276 89027-310 89027-278 89027-312 89027-306 89027-308 89027-262 89027-340 89027-264 89027-260 89027-348 89027-304 89029-312 89027-342 89027-266 89029-314 89027-344 89027-300 89027-268 89027-250 89027-252 89027-296 89027-330 89027-290 89027-292 89027-258 89027-336 89027-338 89027-298 89027-254 89027-332 89029-322 89027-256 89029-324 89027-334 89029-316 89029-318
O-Rings, Perfluoro, Chemglass
Seals Sealing Rings
Perfluoro is a family of perfluoroelastomer compounds designed to provide superior chemical resistance, similar O-rings such as Kalrez® 4079 and Chemraz®.
  • Perfluoro O-rings are stocked in WHITE 75 durometer
  • Effective service temperature is –7 to 230 °C
  • Used with Duran flanges
These O-rings are used with acids, inorganic bases, alcohols/ethers, aromatic hydro-carbons, esters, aldehydes/ ketones, aliphatic hydrocarbons, amines and chlorocarbon solvents.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty