Would you like to log in again?
Wide Mouth Jars with Caps, Polypropylene, Taral® Plastics
Supplier: Taral
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
16-89-WPPWC 1/4-33-WPPWC 1/2-33-WPPWC 4-70-WPPWC 2-53-WPPWC 2-48-WPPWC 8-89-WPPWC 4-53-WPPWC 2-53-NPPWC 8-89-NPPWC 1/2-33-NPPWC 4-70-NPPWC 12-89-WPPWC 1-43-NPPWC 1-38-NPPWC 8-70-WPPWC 32-120-NPPWC 1/4-33-NPPWC 6-70-NPPWC 7/8-33-NPPWC 1-43-WPPWC 6-70-WPPWC 1-38-WPPWC 32-120-WPPWC 12-89-NPPWC
89027-863CS 681.06 USD InStock
89027-863 89027-888 89027-865 89027-882 89027-884 89027-856 89027-858 89027-891 89027-897 89027-874 89027-852 89027-876 89027-898 89027-892 89027-870 89027-895 89027-851 89027-872 89027-904 89027-906 89027-868 89027-900 89027-867 89027-848 89027-902
Wide Mouth Jars with Caps, Polypropylene, Taral® Plastics
Jars General Use Jars
These straight-sided, injection molded plastic jars feature a wide mouth and screw-top
Jars are made of either natural or white polypropylene and are designed with a 400 standard thread neck finish. Natural jars are translucent and white jars are opaque. Jars include a screw-top, ribbed-sided white polypropylene plastic closure with an industry standard 0.030" thick polyethylene foam F-217 liner. All jars are bulk packed in a 2mil polyethylene bag.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty