Would you like to log in again?
Wide Mouth Jars without Caps, Taral® Plastics
Supplier: Taral
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
2-53-CPS 1-43-CPS 12-89-WPP 1-38-NPPC 4-53-NPPC 1/2-33-NPPC 16-89-WPP 16-89-NPPC 8-89-WPP 4-53-CPS 1-43-NPPC 16-89-CPS 4-70-WPP 8-89-NPPC 12-89-NPPC 1/2-33-CPS 16-89-BPP 8-89-NPPC 1-38-WPP 12-89-CPS 7/8-33-UL-CPS 7/8-33-UL-NPPC 2-53-UL-NPPC 4-53-WPP 8-89-CPS 6-70-WPP 1/2-33-WPP 1-43-WPP 6-70-NPP
89027-962CS 390.9 USD InStock
89027-962 89027-940 89027-921 89027-987 89027-961 89027-937 89027-959 89027-918 89027-939 89027-934 89027-999 76102-340 89027-955 89027-974 89027-995 89027-932 89027-976 89027-953 89027-997 89027-991 89027-993 89027-926 89027-989 89027-922 89027-969 89027-925 89028-002 89028-000 89028-004
Wide Mouth Jars without Caps, Taral® Plastics
Jars General Use Jars
These straight-sided, screw-top plastic jars are made of natural, white, or black polypropylene, or clear polystyrene
The ribbed-sided, screw-top caps are made of white polypropylene and include an industry standard 0.030" thick polyethylene foam F-217 liner. Caps are designed with a 400 standard thread finish and are available with a stackable, matted top on sizes 45mm and larger. Caps and jars are sold separately.
Natural jars are translucent and white jars are opaque. Jars are designed with a 400 standard neck thread finish.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty