Would you like to log in again?
Airfree® Schlenk Thermocouple Gauge Adapter, Chemglass
Supplier: Chemglass
Airfree®
AF-0306-03 AF-0306-10
89028-174EA 425.65 USD InStock
89028-174 89028-176
Airfree® Schlenk Thermocouple Gauge Adapter, Chemglass
Adapters Adapters, Joints with Tube Connectors
A (0 to 12 mm) Chem-Vac™ Chem-Cap® valve with an O-Ring joint for connection to the right-hand side of (80064-398) vacuum manifold.
  • Available for #30 O-Ring joints
Delivery information: Complete gauge consists of a glass portion, a stainless steel bushing drilled and tapped for 1/8" NPT, permitting direct connection to gauge tube, and an open top GL-25 screw cap.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty