Would you like to log in again?
Oil for Welch® Vacuum Pumps, Chemglass
Supplier: Chemglass
AF-0360-12 AF-0360-13 AF-0360-16 AF-0360-10 AF-0360-14 AF-0360-17 AF-0360-11
89028-182EA 647.01 USD InStock
89028-182 89028-192 89028-180 89028-190 89028-186 89028-184 89028-178
Oil for Welch® Vacuum Pumps, Chemglass
Lubricants
Oil is specially formulated for use in either belt driven or direct drive vacuum pumps (AF-0349 or AF-0350)
  • Oil is available in three types, Duo-Seal®, Premium and Gold
The Duo-Seal oil is for use with belt driven (low RPM) AF-0350 pumps.
The Premium oil is a triple distilled hydrocarbon oil resistant to breakdown at the high RPM of direct drive pumps.
The Gold oil is a double distilled synthetic hydrocarbon oil that has no aromatic compounds or sulfur. Gold oil will offer longer life and superior protection for the internal components in corrosive pumping applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty