Would you like to log in again?
Thermocouple Adapters, PTFE, with Perfluoro O-Ring, Chemglass
Supplier: Chemglass
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
CG-1048-P-07 CG-1048-P-03 CG-1048-P-02 CG-1048-P-05 CG-1048-P-04 CG-1048-P-01 CG-1048-P-06
89028-230EA 129.65 USD InStock
89028-230 89028-240 89028-234 89028-232 89028-242 89028-238 89028-236
Thermocouple Adapters, PTFE, with Perfluoro O-Ring, Chemglass
Adapters Adapters, Conical Ground Joints
PTFE adapter for use with either 1/8" or 1/4" thermocouples.
  • Outer perfluoro O-Ring forms a grease-free, airtight seal between the adapter and the listed size of outer joint
  • Adapter design permits use with Keck clips
A compression nut works in conjunction with the upper perfluoro O-Ring to provide an airtight seal between the adapter and thermocouple.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty