Would you like to log in again?
Moisture Test Adapters, Chemglass
Supplier: Chemglass
CG-1063-04 CG-1063-09 CG-1063-05 CG-1063-01
89028-292EA 354.09 USD InStock
89028-292 89028-290 89028-284 89028-300
Moisture Test Adapters, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters are used to construct a simple moisture trap.
Adapters with larger center to center spacing can be used with process reactors and larger flasks up to 50 l. Add a condenser to the top outer joint and a graduated funnel to the lower inner joint.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty