Would you like to log in again?
Glassware Cleaning Brushes for Cylindrical Reactors, Chemglass
Supplier: Chemglass
CG-1146-01 CG-1146-02
89028-394EA 32 USD InStock
89028-394 89028-392
Glassware Cleaning Brushes for Cylindrical Reactors, Chemglass
Brushes
Glassware cleaning brush for large cylindrical reactors.
  • Chemically resistant brush head
  • Will not scratch surfaces
Brush has a 3×4.5" rectangular non-abrasive scrubbing pad at the end of either a 9" or 33" handle for cleaning larger cylindrical reactors.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty