Would you like to log in again?
Bottle Cap Assembly, Chemglass
Supplier: Chemglass
CG-1158-15 CG-1158-12
89028-398EA 100.88 USD InStock
89028-398 89028-400
Bottle Cap Assembly, Chemglass
Closures Bottle Closures
Bottle cap for GL-45 and GPI-38 threads that permits standard solvent or media bottles to be used as an HPLC reservoir.
  • Caps are made from polypropylene
  • Inserts are made from tefzel
Complete cap assemblies consists of the cap, tefzel insert, two plugs for the 1/8" tubing openings and a plug for the opening. Caps have two holes to accomodate 1/8" tubing and one hole that accepts a standard luer inner joint.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty