Would you like to log in again?
Adapters, Rotary Evaporators, Vials, Chemglass
Supplier: Chemglass
CG-1318-41 CG-1318-42
89028-424EA 291.97 USD InStock
89028-424 89028-422
Adapters, Rotary Evaporators, Vials, Chemglass
Adapters Adapters, Conical Ground Joints
Adapters permit the connection of standard laboratory dram sample vials directly to the vapor tube or a bump trap of a rotary evaporator.
  • Adapters feature a top outer joint
Adapters have a top outer joint of the listed size and a 20 to 400 G.P.I. thread at the bottom. A PTFE-lined, polypropylene, double-threaded connector attaches the vial to the adapter.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty