Would you like to log in again?
ECO Funnels with HPLC Waste Hook Adapter, Chemglass
Supplier: Chemglass
CG-1759-A-01 CG-1759-A-06
89028-712EA 291.16 USD InStock
89028-712 89028-720
ECO Funnels with HPLC Waste Hook Adapter, Chemglass
Funnels Safety Funnels
ECO funnels are supplied with an HPLC waste hook adapter that connects to the side of the funnel, eliminating the danger of dangling waste lines into bottles and containers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty