Would you like to log in again?
Ribbed, Standard Stem Funnels, Chemglass
Supplier: Chemglass
CG-1770-02 CG-1770-01 CG-1770-03 CG-1770-04
89028-800EA 67 USD InStock
89028-800 89028-794 89028-798 89028-796
Ribbed, Standard Stem Funnels, Chemglass
Funnels Filtration Funnels Filter Funnels
Polypropylene funnels feature the exclusive filter-step internal design for speedier filtrations. External ribs prevent airlock.
  • Parallel stem measures 8 mm O-Ø
  • Steam autoclavable at 121 °C (250 °F)
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty